Learn More
Medchemexpress LLC Ibutamoren mesylate | 159752-10-0 | MFCD00942284 | >98.0% | 624.77 | C28H40N4O8S2 | 10 MG

Supplier: Medchemexpress LLC HY5084410MG
Ibutamoren mesylate (MK-677) is an orally active, non-peptide growth hormone secretagogue receptor (GHSR) agonist supplied as the mesylate salt for laboratory research. It is used in pharmacological and endocrine studies to elevate growth hormone and IGF-1 levels in preclinical models.
- Orally active GHSR agonist for growth hormone and IGF-1 studies.
- Mesylate salt form with molecular formula C28H40N4O8S2 and molecular weight 624.77.
- Available in 1 mg, 5 mg, and 10 mg packages for small-scale laboratory use.
- Purity typically ≥98% as supplied by reputable distributors.
- Storage: sealed at 4°C; in solvent store at -80°C up to 6 months or -20°C up to 1 month.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.