Learn More
Medchemexpress LLC Ibutilide | 122647-31-8 | 99.0% | 5 MG

Supplier: Medchemexpress LLC HYB0387A5MG
Ibutilide (U70226E free base) is an action potential-prolonging antiarrhythmic agent that acts as a potent blocker of the rapidly activating delayed rectifier K+ current (IKr) in AT-1 cells. It inhibits Cav1.2 current in CHO cells and prolongs cardiac repolarization in vivo.
- Potent IKr blocker with an EC50 of 20 nM at +20 mV in atrial tumor myocytes.
- Inhibits Cav1.2 current with an IC50 of 62.5 μM in CHO cells.
- Blocks IKr in HERG+MDR1*1 and HERG alone, showing resistance in MDR1*7 expressing cells.
- Store pure form at -20°C for 3 years or 4°C for 2 years.
- Store in solvent at -80°C for 6 months or -20°C for 1 month.
- Shipping at room temperature if less than 2 weeks.
- Keep container sealed in a cool, well-ventilated area, away from direct sunlight and ignition sources.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.