Learn More
AdipoGen IBX pyridine complex
SDP
Supplier:  AdipoGen CDXH0104G005
Encompass_Preferred
Chemical. CAS 1380548-11-7. Formula C12H10INO4. MW 359.1. Synthetic. Stable pyridinium salt of o-iodoxybenzoic acid PIBX is a convenient substitute of o-iodoxybenzoic acid IBX as an oxidant. PIBX is safer, has neutral properties behaves as an equivalent to IBX in the oxidation of alcohols to ketones or aldehydes in polar solvents and provides higher oxidation rate in THF due to better solubility.
Catalog No. 50-256-0728