Learn More
Selleck Chemical LLC IC-87114 5mg 371242-69-2
SDP
Supplier:  Selleck Chemical LLC S12685MG
Encompass_Preferred
IC-87114 is a selective PI3K? inhibitor with IC50 of 0.5 ?M in a cell-free assay, 58-fold more selective for PI3K? than PI3K?, and over 100-fold more selective than PI3K?/?. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-856-8