Learn More
SPEX CERTIPREP LLC 3P IC-BR 100 UG/ML BROMIDE IN H2O
Supplier:  SPEX CERTIPREP LLC 3P ICBR100
Encompass_Preferred
(HPS by Zeptometrix) Bromide at 100 g/mL in H2OThis standard is traceable to NIST SRM 3184 and certified ISO Guide 34 and 17025.18 Month Expiry DateSingle Component Ion Chromatography Standard
Catalog No. NC1446395