Learn More
Thermo Scientific IC TUBING 1INCH/EACH
Supplier:  Thermo Scientific 044221
Encompass
Tubing, Color: Red, Size: 0.005 in. I.D., Volume: 0.004mL/ ft, Back Pressure: 111.360psi/ ft at 1mL/min.
Catalog No. NC9179171