Learn More
Medchemexpress LLC Ica 110381 | 325457-99-6 | 99.8% | 267.11 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY108587
Encompass_Preferred
ICA 110381 is an orally active Kv7.2/Kv7.3 (KCNQ2/3) potassium channel activator with anticonvulsive properties. It is a KCNQ2/Q3 activator (EC50=0.38 μM) and a KCNQ1 antagonist (IC50=15 μM). It induces rubidium efflux in CHO cells expressing KCNQ2 or KCNQ3, and inhibits rubidium efflux from CHO cells expressing KCNQ1.
  • Increases M-currents (IM) in CA1 pyramidal cells
  • Damps neuronal excitability
  • Elevates the threshold for induction of afterdischarges in animal studies
  • Reduces seizure severity and duration in animal studies
Catalog No. 50-003-47416