Learn More
Medchemexpress LLC ICAM-1-IN-1 25mg | 25mg
SDP
Supplier:  Medchemexpress LLC HYU0000325MG
Encompass_Preferred
ICAM-1-IN-1 is a potent and selective inhibitor of E-selectin and ICAM-1 with IC50 values of 7 and 5 nM respectively
Catalog No. 50-003-22195