Learn More
Medchemexpress LLC Icapamespib hydrochloride | 2267287-26-1 | 98.7% | 599.32 g/mol | C19H25Cl2IN6O2S | 5 MG

Supplier: Medchemexpress LLC HY137441A5MG
Icapamespib hydrochloride is the hydrochloride salt of icapamespib, a selective, orally active inhibitor of epichaperomes assembled by HSP90. Supplied as a high-purity solid for research use, it is intended for biochemical and cellular studies of HSP90-related epichaperome biology and therapeutic modulation.
- Selective inhibitor of epichaperomes assembled by HSP90.
- Intended for biochemical and cellular research applications.
- High purity: 98.7%.
- Molecular weight: 599.32 g/mol.
- Chemical formula: C19H25Cl2IN6O2S.
- White to off-white solid appearance.
- Recommended storage sealed and away from moisture; in solvent: -80°C (6 months), -20°C (1 month).
- Available in 5 mg packaging.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.