Learn More
PROTEIN CAPTURE SCIENCE LLC ICAPTAG BULK RESIN 10ML

Supplier: PROTEIN CAPTURE SCIENCE LLC 05001002
- Advanced affinity chromatography resin
- Self-cleaving affinity tag system for tagless proteins
- Compatible with various expression systems including E. coli, CHO, and HEK
- Strong and selective binding at pH 8.5
- Tag released with high purity and yield at pH 6.2
- Cleavage completion in less than 5 hours
- Resin matrix made of crosslinked agarose
- Compatibility with salts, sugars, and protease inhibitors
- Regenerable with low pH wash at pH ≤ 2.0
- Highly reusable, cost-effective for biomanufacturing
- Volume of 10 mL
- Ethanol content of 18% ± 2%
- Store at 2-8 °C
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.