Learn More
TARGETMOL CHEMICALS INC ICARITIN 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T339810MG
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Icaritin (Anhydroicaritin) has hormone regulation activity and cardiovascular function improvement activity. Icaritin has anticancer activity can induce S phase arrest and apoptosis inhibit ENKL cell proliferation. Icaritin has anti-multiple myeloma activity mainly mediated by inhibiting IL-6/JAK2/STAT3 signaling. Icaritin at low concentration (4 or 8 uMol/L) can promote rat chondrocyte proliferation and inhibit cell apoptosis while the effect of Icaritin on rat chondrocyte at high concentration was reversed. purity: 98%
Catalog No. 50-257-6950