Learn More
Medchemexpress LLC Icatibant acetate | 138614-30-9 | 99.9% | 1364.57 g·mol⁻¹ | C61H93N19O15S | 5 MG

Supplier: Medchemexpress LLC HY1088965MG
Icatibant acetate is a synthetic peptide antagonist of the bradykinin B2 receptor used in pharmacology and signaling research. Supplied as the acetate salt, it is intended for in vitro and biochemical studies requiring a selective B2 antagonist.
- Potent bradykinin B2 receptor antagonist (IC50 1.07 nM; Ki 0.798 nM).
- Peptide sequence appropriate for receptor pharmacology studies.
- High purity (99.85%) suitable for sensitive assays.
- Molecular weight 1364.57 g·mol⁻¹ for accurate dosing.
- Available in multiple milligram sizes for dose-ranging experiments.
- Provided as an acetate salt to improve compound stability.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.