Learn More
Medchemexpress LLC HY-108896 1mg Medchemexpress, Icatibant (acetate) CAS:138614-30-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1088961MG
Encompass_Preferred
Medchemexpress, HY-108896 1mg Icatibant (acetate) CAS:138614-30-9 Icatibant acetate (HOE-140 acetate) is a potent and specific peptide antagonist of bradykinin B2 receptor with IC50 and Ki of 1.07 nM and 0.798 nM respectively[2][3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9650