Learn More
Cellular Dynamics International iCell DopaNeurons PD SNCA A53T, iPSC-derived dopaminergic neurons, Heterozygous SNCA A53T mutation, Cryopreserved format, ≥5.0 x 10^6 viable cells
Supplier:  Cellular Dynamics International R1111
Encompass
iCell DopaNeurons PD SNCA A53T, iPSC-derived dopaminergic neurons, Heterozygous SNCA A53T mutation, Cryopreserved format, ≥5.0 x 10^6 viable cells
Catalog No. NC3363434