Learn More
Cellular Dynamics International iCell Microglia AD TREM2, 1 x 10^6 cells per vial, Cryopreserved, TREM2 Homozygous Knockout, Human, Male, Age 50-59
Supplier:  Cellular Dynamics International R1202
Encompass
iCell Microglia AD TREM2, 1 x 10^6 cells per vial, Cryopreserved, TREM2 Homozygous Knockout, Human, Male, Age 50-59
Catalog No. NC3251630