Learn More
Medchemexpress LLC Icg maleimide | 2143933-81-5 | 96.4% | 853.08 | C51H56N4O6S | 10 MG
SDP
Supplier:  Medchemexpress LLC HYD174410MG
Encompass_Preferred
ICG Maleimide is a thiol-reactive near-infrared (NIR) fluorescent dye used to label thiol-containing biomolecules and generate a stable NIR fluorescence signal for bioimaging applications (Ex/Em 789/813 nm). It is supplied as a solid, green to dark green material with high purity suitable for conjugation to proteins and peptides.
  • Thiol-reactive maleimide functional group for selective labeling of cysteine residues.
  • Near-infrared excitation/emission at 789/813 nm for deep tissue imaging.
  • High purity suitable for research conjugation and imaging applications.
  • Solid form, green to dark green color, easy to handle and store.
  • Storage recommendations: protect from light; in solvent, -80°C for 6 months and -20°C for 1 month.
Catalog No. 50-002-97461