Learn More
Abcam ICI 182,780, Estrogen receptor antagonist, 50MG
Supplier:  Abcam AB12013150MG
Encompass_Preferred
MW 606.8 Da, Purity >99%. Estrogen receptor antagonist with no partial agonist activity. Anticancer agent (IC₅₀ = 0.29 nM for inhibition of breast cancer cell growth). Causes downregulation and loss of estrogen receptors.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7859