Learn More
METTLER TOLEDO iControl - Single Instrument License
Supplier:  METTLER TOLEDO 51161893
Encompass
iControl™ enables chemists and engineers to gain an in-depth understanding of the reaction data collected by EasyMax™/OptiMax™/RC1mx™/RX-10™. The simple graphical interface helps users quickly identify key reaction events and parameters to compare data from multiple experiments. iControl™ can be used either in combination with the EasyMax™/OptiMax™/RC1mx™/RX-10™ touchpad or on its own to create and control experiments.
Catalog No. NC1331323