Learn More
Medchemexpress LLC Icotinib (Hydrochloride) | 1204313-51-8 | C22H22ClN3O4 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY15164100MG
Encompass_Preferred
Icotinib Hydrochloride (BPI-2009) is a potent, CNS-penetrant, and specific EGFR inhibitor with an IC50 of 5 nM. It inhibits mutant EGFRL858R, EGFRL858R/T790M, EGFRT790M, and EGFRL861Q. This compound is a click chemistry reagent containing an Alkyne group, enabling it to undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • Potent, CNS-penetrant, and specific EGFR inhibitor
  • Inhibits mutant EGFRL858R, EGFRL858R/T790M, EGFRT790M, and EGFRL861Q
  • Click chemistry reagent for copper-catalyzed azide-alkyne cycloaddition
  • Exhibits potent dose-dependent antitumor effects in nude mice
  • Well tolerated at doses up to 120 mg/kg/day in mice
Catalog No. 50-003-73933