Learn More
Inorganic Ventures 10µg/mL LEAD 125mL
SDP
Supplier:  Inorganic Ventures MSPB10PPM125ML
Encompass_Preferred
Matrix: Nitric Acid (HNO3), Starting Material: Pb(NO3)2, Analyte: Pb 10 ug/mL.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search
Catalog No. 50-723-756