Learn More
Perkin Elmer US LLC ICP-OES Oil-Less Air Compressor with Dryer in Silencing Cabinet, 220 V/60 Hz
Supplier:  Perkin Elmer US LLC N0777607
Encompass_Preferred
One version includes a silencing cabinet resulting in a compressor operating at a very quiet noise level of 55 dB/A. All models are equipped with ASME rated pressure tanks and safety valves, line pressure regulator, gauges and line ball valve for a complete turnkey compressor station. Horse Power: 1.5 HP Output CFM/Output L: 6.0 CFM/170 L/min Max Pressure PSI/Max Pressure Bar: 120 PSI/8 bar Noise Level: 72 dB/A, with Enclosure 55 dB/A Tank Size Gal./Tank Size L: 13 Gal./50 L Dimensions without Enclosure: 25 in. x 18 in. x 29 in. Dimensions with Enclosure: 29 in. x 23 in. x 32 in. Weight without Enclosure: 97 lbs. Weight with Enclosure: 207 lbs. Packed Dimensions without Enclosure: 28 in. x 21 in. x 29 in. Packed Dimensions with Enclosure: 33 in. x 28 in. x 39 in
Catalog No. 50-210-4205
Only null left
Add to Cart
Request a Quote