Learn More
Apexbio Technology LLC Idarubicin HCl 57852-57-0 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC A24765.1
Encompass_Preferred
Idarubicin hydrochloride an anthracycline derivative structurally related to daunorubicin acts primarily by inhibiting DNA topoisomerase II It exerts cytotoxic effects through stabilization of DNA-topoisomerase II intercalation complexes inducing DNA strand breaks and apoptosis Idarubicin can be enzymatically activated intracellularly via NADPH-cytochrome C reductase enhancing DNA damage Experimentally it is commonly utilized in leukemia research particularly acute myeloid leukemia (AML) assessing cellular response to DNA damage apoptosis induction and mechanisms underlying drug resistance Idarubicin exhibits IC50 values typically ranging from low nanomolar to micromolar concentrations depending on cell line and assay conditions
Catalog No. 50-000-62976