Learn More
Medchemexpress LLC Idarubicin (hydrochloride) | 57852-57-0 | 99.9% | 533.95 | 1 ML
Supplier: Medchemexpress LLC HY1738110MM/1ML
Idarubicin hydrochloride is an anthracycline antileukemic agent that inhibits topoisomerase II, interfering with DNA replication and RNA transcription. It also inhibits the growth of bacteria and yeasts, and is intended for research use only.
- Inhibits DNA replication
- Interferes with RNA transcription
- Active against bacteria and yeasts
- Greater cytotoxicity than daunorubicin or doxorubicin in various in vitro systems