Learn More
Cayman Chemical IdarubIcInhydrochlorIde 10mg
SDP
Supplier:  Cayman Chemical 1417610 MG
Encompass_Preferred
A 4-demethoxy analog of daunorubicin which intercalates in DNA and inhibits topoisomerase II, resulting in cancer cell cytotoxicity at low concentrations (IC50 = 20-120 nM); effective in combination therapy for the treatment of different types of leukemia
Catalog No. 50-295-3477