Learn More
Medchemexpress LLC Idasanutlin-d3-1 (RG7388-d3-1) | C31H26D3Cl2F2N3O4 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY153939S1MG
Encompass_Preferred
Idasanutlin-d3-1 (RG7388-d3-1) is the deuterium-labeled form of Idasanutlin, a potent antagonist of MDM2/p53. It has been shown to inhibit relapsed or refractory acute myeloid leukemia.
  • Deuterium-labeled compound.
  • Stable heavy isotopes serve as tracers for quantitation during drug development.
  • Deuteration can affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-003-76782