Learn More
Selleck Chemical LLC Idasanutlin (RG-7388) 5mg 1229705-06-9
SDP
Supplier:  Selleck Chemical LLC S72055MG
Encompass_Preferred
Idasanutlin (RG-7388) is a potent and selective p53-MDM2 inhibitor with IC50 of 6 nM showing improved in vitro binding as well as cellular potency/selectivity. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-4515