Learn More
BIOGEMS INTERNATIONAL INC IDE1 10mg
SDP
Supplier:  BIOGEMS INTERNATIONAL INC 116489910MG
Encompass
IDE1 activates the TGF-beta signaling pathway and induces definitive endoderm formation in human and mouse embryonic stem cells. It is also reported to increase the levels of Nodal expression, induce Smad2 phosphorylation and SOX17 expression.
Catalog No. 50-152-9665