Learn More
STEMCELL Technologies IDE1, Size: 5 mg
SDP
Supplier:  STEMCELL Technologies 72514
Encompass
Activate the Activin/BMP/TGF-β pathway with inducer of definitive endoderm 1 (IDE1; Borowiak et al. Cell Stem Cell, 2009). This small molecule induces the differentiation of mouse or human embryonic stem (ES) cells to definitive endoderm in the absence of Activin A, NODAL, or feeder cells (Borowiak et al. Cell Stem Cell, 2009), and the differentiation of mouse or human pluripotent stem cells by activating SMAD2 phosphorylation and NODAL expression (Borowiak et al. Cell Stem Cell, 2009). At EC₅₀ = 125 nM, SOX17 expression was induced in mouse ES cells. IDE1 is supplied as a crystalline solid with ≥95% purity; Molecular weight 306.3 g/mol; CAS Number 1160927-48-9. Visit STEMCELL.com for more information and related products.
Catalog No. 50-197-5934