Learn More
Thomas Scientific Identi-Marker, Fine Tip, Green
SDP
Supplier:  Thomas Scientific 1201U07
Encompass_Preferred
Writes on foam, plastic, glass and metal Ideal for use with Identi-Plug s® and other Jaece products. Writes on cold or wet surfaces. Marks on plastic, glass and metal. Ideal for general laboratory, hospital or industrial use. Unique Features Economical Permanent, quick-drying ink Extra-fine, durable plastic tip Alcohol resistant
Catalog No. NC1357401