Learn More
Cole-Parmer Idex 0001-6354 Vacuum Degasser, 4 Channel, 480 µL Per Channel, 2 mL/min; 100 to 240 VAC
Supplier:  Cole-Parmer 0201294
Encompass
These in-line full stand-alone degassing systems efficiently remove unwanted dissolved gases from mobile phases. Their unique design provides the highest level of reliable performance and makes helium sparging unnecessary. Optimal continuous operation is maintained by vacuum system monitoring.
Catalog No. NC1630962