Learn More
Trajan Scientific Idex P-137X Super Flangeless Nut Peek | Standard Knurl | 3/16" OD Tubing | 5/16-24 Flat Bottom | 10/PK
Supplier:  Trajan Scientific ID.P137X
Encompass
Combine with Super Flangeless ferrule for a high pressure holding flat-bottom fitting assembly. It Eliminates tubing twist that can loosen connections. It is resistant to lossening through vibration.
Catalog No. 50-187-1406