Learn More
Selleck Chemical LLC IDF-11774 S8771-10MM/1ML
SDP
Supplier:  Selleck Chemical LLC S877110MM/1ML
Encompass_Preferred
IDF-11774 is a hypoxia-inducible factor-1 (HIF-1) inhibitor It reduces the HRE-luciferase activity of HIF-1 (IC50 3 65 M) and blocks HIF-1 accumulation under hypoxia in HCT116 human colon cancer cells
Catalog No. 50-000-24498