Learn More
Scytek Laboratories Anti-IDH1 R132H Antibody, High Specificity for Tumor Detection, Mouse Monoclonal, 0.5 ml
SDP
Supplier:  Scytek Laboratories DIAH09
Encompass
The SCYTEK LABORATORIES IDH1 R132H Antibody is a highly specific mouse monoclonal antibody designed for detecting the IDH1 R132H point mutation in human tissues. It is essential for the differential diagnosis of gliomas, aiding in the classification of astrocytomas and oligodendrogliomas. This antibody aids in research applications by allowing identification of single tumor cells and supporting prognosis assessments for IDH1-mutated tumors, distinguishing them from wild-type counterparts.
Catalog No. NC1603148