Learn More
Scytek Laboratories IDH1 R132H; Clone H09 (Concentrate), 0.1mL
SDP
Supplier:  Scytek Laboratories DIAH09M
Encompass
Species: Mouse, Immunogen: Synthetic peptide, amino acid sequence CKPIIIGHHAYGD, Clone: H09, Isotype: IgG2a, Species Reactivity: Human, Positive Control: Oligodendroglioma, diffuse astrocytoma, Specificity: Human IDH1 R132H point mutation
Catalog No. NC1632336