Learn More
AdipoGen anti-Isocitrate Dehydrogenase 1 [IDH1] (R132H Mutant) (human), mAb (AG-IHC132) (100 µl)
SDP
Supplier:  AdipoGen AG20B6024R100
Encompass_Preferred
Monoclonal Antibody. Recognizes human IDH1 mutated at R132H. Applications: ELISA, IHC. Isotype: Mouse IgG1. Clone: AG-IHC132. Liquid. In Tris Buffer, pH 7.4, containing 1% BSA and <0.1% sodium azide. Isocitrate Dehydrogenase 1 (IDH1) is a soluble, cytosolic enzyme involved in the TCA metabolic cycle. The most notable mutation in this enzyme, R132H, is clinically indicated in the majority of astrocytomas and oligodendroglial tumors, with the mutation being associated with more favourable prognosis and increased survival in those patients. IDH1 R132H is also useful in the differential diagnosis between anaplastic glioma and glioblastoma.This antibody is intended to qualitatively identify by light microscopy the presence of associated antigens in sections of formalin-fixed, paraffin-embedded tissue sections using IHC test methods. It has been optimized and validated using the BOND-MAX fully automated IHC&ISH stainer (see Protocol).
Catalog No. 50-205-3009