Learn More
Medchemexpress LLC IDO-1 Mouse His 500ug
SDP
Supplier:  Medchemexpress LLC HYP72616500UG
Encompass_Preferred
IDO Protein an enzyme is involved in the metabolism of tryptophan Dysregulation of IDO Protein has been linked to immune escape mechanisms and tumor progression Targeting IDO Protein may provide potential therapeutic interventions in cancer by modulating tryptophan metabolism enhancing immune responses and inhibiting tumor growth IDO-1 Protein Mouse (His) is the recombinant mouse-derived IDO protein expressed by E coli with N-6 His labeled tag
Catalog No. 50-001-82085