Learn More
Medchemexpress LLC IDO-2 Human His 500ug

Supplier: Medchemexpress LLC HYP70863500UG
Recombinant human IDO-2 protein with a C-terminal 6xHis tag, expressed in E. coli and supplied as a 0.22 μm-filtered PBS solution. The protein is purified to greater than 95% by reducing SDS-PAGE, provided at approximately 0.707 mg/mL, and is suitable for biochemical assays, antibody development, and structural studies. Store frozen and avoid repeated freeze-thaw cycles.
- Purity greater than 95% as determined by reducing SDS-PAGE.
- C-terminal 6xHis tag for affinity purification and detection.
- Expressed in E. coli for efficient recombinant production.
- Supplied in PBS, pH 7.4, filtered through 0.22 μm.
- Provided at ~0.707 mg/mL concentration; 500 μg format available.
- Store at -80°C for long-term stability; avoid repeated freeze-thaw.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.