Learn More
Medchemexpress LLC Ido-in-1 | 914638-30-5 | 98.1% | C9H7BrFN5O2 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY7953110MM/1ML
Encompass_Preferred
IDO-IN-1 is a potent indoleamine 2,3-dioxygenase (IDO) inhibitor. It has an IC50 of 59 nM and a HeLa IC50 of 12 nM.
  • Potent indoleamine 2,3-dioxygenase (IDO) inhibitor
  • IC50 of 59 nM
  • HeLa IC50 of 12 nM
  • For research use only
Catalog No. 50-004-33174