Learn More
Medchemexpress LLC HY-115122 1mg Medchemexpress, IDO-IN-12 CAS:1888341-29-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1151221MG
Encompass_Preferred
Medchemexpress, HY-115122 1mg IDO-IN-12 CAS:1888341-29-4 IDO-IN-12 is an indoleamine 2,3-dioxygenase (IDO) inhibitor extracted from patent WO 2017181849 A1. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9378