Learn More
Apexbio Technology LLC IDO-IN-2 1668565-74-9 5mg
SDP
Supplier:  Apexbio Technology LLC B83105
Encompass_Preferred
IDO-IN-2 (CAS 1668565-74-9) is a small-molecule inhibitor of indoleamine 2 3-dioxygenase (IDO) an enzyme involved in the catabolism of tryptophan In vitro studies indicate that IDO-IN-2 efficiently attenuates IDO activity in HeLa cells as evidenced by low IC50 values Preclinical investigations suggest that the compound can modulate synaptic plasticity within regions such as the anterior cingulate cortex and amygdala contributing to the alleviation of neuropathic pain and related symptoms IDO-IN-2 serves as a valuable tool for research in neuropharmacology and cancer immunotherapy facilitating the study of IDO pathway modulation
Catalog No. 50-000-88046