Learn More
Medchemexpress LLC HY-18770 5mg Medchemexpress, IDO-IN-5 CAS:1402837-79-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY187705MG
Encompass_Preferred
Medchemexpress, HY-18770 5mg IDO-IN-5 CAS:1402837-79-9 IDO-IN-5 (NLG-1489) is an indoleamine 2,3-dioxygenase ( IDO ) inhibitor extracted from patent WO WO2012142237A1, compound 1489, has an IC 50 of 1-10 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2820