Learn More
Medchemexpress LLC HY-13983 10mg Medchemexpress, IDO-IN-7 CAS:1402836-58-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1398310MG
Encompass_Preferred
Medchemexpress, HY-13983 10mg IDO-IN-7 CAS:1402836-58-1 IDO-IN-7 (NLG-919 analogue) is a a potent IDO1 inhibitor with an IC50 of 38 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0500
Due to product restrictions, please to purchase or view availability for this product.