Learn More
Medchemexpress LLC Idoxuridine | 54-42-2 | 99.8% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB030710MM/1ML
Encompass_Preferred
Idoxuridine is an iodinated thymidine analogue that acts as a competitive inhibitor of phosphorylases. It inhibits viral activity, particularly viral eye infections like herpes simplex keratitis, by inhibiting DNA polymerase and affecting viral replication. It demonstrates an IC50 value of 4.3 μM against feline herpesvirus and shows anti-orthopoxvirus activity.
  • Inhibits viral activity
  • Acts as a competitive inhibitor of phosphorylases
  • Inhibits DNA polymerase
  • Affects viral replication
  • Shows anti-orthopoxvirus activity
Catalog No. 50-004-05470