Learn More
Medchemexpress LLC Idoxuridine | 54-42-2 | 99.95% | 10 G
SDP
Supplier:  Medchemexpress LLC HYB030710G
Encompass_Preferred
Idoxuridine is an iodinated thymidine analogue that competitively inhibits phosphorylases. It demonstrates viral inhibitory activity, particularly against viral eye infections such as herpes simplex keratitis, by inhibiting DNA polymerase and disrupting viral replication.
  • Competitively inhibits phosphorylases
  • Inhibits viral activity, including herpes simplex keratitis
  • Acts by inhibiting DNA polymerase
  • Affects viral replication
  • Displays anti-orthopoxvirus activity
  • Can stimulate the production of hemolysin plaque-forming cells to sheep red blood cells in animal models
Catalog No. 50-004-06357