Learn More
Medchemexpress LLC Idoxuridine | 54-42-2 | 99.95% | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYB0307
Encompass_Preferred
Idoxuridine (5-Iodo-2′-deoxyuridine) is an iodinated thymidine analogue that competitively inhibits phosphorylases. It can inhibit viral activity, particularly viral eye infections, including herpes simplex keratitis, by inhibiting DNA polymerase and affecting viral replication. It has an IC50 value of 4.3 μM against feline herpesvirus and shows anti-orthopoxvirus activity.
  • Competitively inhibits phosphorylases
  • Inhibits viral activity, especially viral eye infections
  • Inhibits DNA polymerase
  • Affects viral replication
  • Has an IC50 value of 4.3 μM against feline herpesvirus
  • Shows anti-orthopoxvirus activity
Catalog No. 50-004-07074