Learn More
Selleck Chemical LLC Idoxuridine S1883-1g
SDP
Supplier:  Selleck Chemical LLC S18831G
Encompass_Preferred
Idoxuridine (NSC 39661 SKF 14287 5-Iodo-2 -deoxyuridine 5-IUdR IdUrd) is an antiviral agent for feline herpesvirus type-1 with IC50 of 4 3 M
Catalog No. 50-311-0542