Learn More
Selleck Chemical LLC IDRA-21 S2434-25mg
SDP
Supplier:  Selleck Chemical LLC S243425MG
Encompass_Preferred
IDRA-21 is a partial allosteric modulator of AMPA receptor desensitization IDRA-21 is a potent cognition-enhancing drug virtually devoid of neurotoxic liability
Catalog No. 50-311-2692