Learn More
Integrated DNA Technologies Inc 1 Liter IDTE pH 8.0 (1X TE Solution)
Supplier:  Integrated DNA Technologies Inc 11050109
Encompass
IDTE (10 mM Tris, 0.1 mM EDTA) is our recommended solution for resuspending and storing single-stranded DNA and RNA oligos. It offers the most stability for the longest duration when compared to oligos stored dry or in water. IDTE is available at pH 7.5 or pH 8.0.
Catalog No. NC0589807
This product needs to be quoted. Please contact your Fisher Scientific Sales Representative for pricing.
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.