Learn More
Medchemexpress LLC iE-DAP dihydrochloride | 99.7% | 392.23 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYP5523A1MG
Encompass_Preferred
iE-DAP dihydrochloride is a Nod1 agonist. It activates the NOD1 receptor, triggering NF-κB and MLCK signaling pathways, which lead to cellular inflammatory responses and tight junction disruption. It downregulates ZO-1 and Occludin genes. In human trophoblast cell cultures, it increases secretion of cytokines like IL-6, GRO-α, MCP-1, IL-8, and MIP-1β. In pregnant mice, iE-DAP dihydrochloride induces preterm birth, reduces fetal body weight, and causes fetal inflammation, making it applicable for studies related to mastitis and preterm birth.
  • Nod1 agonist.
  • Activates NF-κB and MLCK signaling pathways.
  • Downregulates ZO-1 and Occludin genes.
  • Increases secretion of inflammatory cytokines.
  • Induces preterm birth in pregnant mice.
  • Relevant for mastitis and preterm birth studies.
Catalog No. 50-003-96995