Learn More
METASPEC IEC 62535 Copper Paper Catalyst Test Pieces - copper wrapped in kraft paper per IEC 62535, sold in multiples of 20
Supplier:  METASPEC 15311062535
Encompass
Copper test specimens for corrosion testing per IEC 62535. These copper test strips are made from EN 13601 / UNS C11000 copper, and are 7.5 mm x 30 mm x 1.5 mm (0.3" x 1.2" x 1/16"). Each copper strip is wrapped in four layers of IEC 60554-3-1 kraft paper and is individually packaged. Sold in multiples of 20.
Catalog No. NC3959118